Skip to Content
Merck
All Photos(1)

Documents

G3278

Sigma-Aldrich

Geranylgeraniol

≥85% (GC)

Synonym(s):

all trans-3,7,11-15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O
CAS Number:
Molecular Weight:
290.48
Beilstein:
1913779
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥85% (GC)

form

liquid

solubility

ethanol: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CO

InChI

1S/C20H34O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h9,11,13,15,21H,6-8,10,12,14,16H2,1-5H3/b18-11+,19-13+,20-15+

InChI key

OJISWRZIEWCUBN-QIRCYJPOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

As the pyrophosphate, the metabolic precursor of all diterpenes. Screening in vitro shows this to be a potent and selective agent against Mycobacterium tuberculosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miu Iijima et al.
Plant physiology, 174(4), 2213-2230 (2017-07-07)
Daurichromenic acid (DCA) synthase catalyzes the oxidative cyclization of grifolic acid to produce DCA, an anti-HIV meroterpenoid isolated from
Kenro Tokuhiro et al.
Applied and environmental microbiology, 75(17), 5536-5543 (2009-07-14)
(E, E, E)-Geranylgeraniol (GGOH) is a valuable starting material for perfumes and pharmaceutical products. In the yeast Saccharomyces cerevisiae, GGOH is synthesized from the end products of the mevalonate pathway through the sequential reactions of farnesyl diphosphate synthetase (encoded by
Aida Hiseni et al.
Applied microbiology and biotechnology, 91(4), 1029-1036 (2011-05-19)
Two carotenoid 1,2-hydratase (CrtC) genes from the photosynthetic bacteria Rubrivivax gelatinosus and Thiocapsa roseopersicina were cloned and expressed in Escherichia coli in an active form and purified by affinity chromatography. The biochemical properties of the recombinant enzymes and their substrate
Jianzhong Wen et al.
Biochemistry, 49(26), 5455-5463 (2010-06-05)
The Fenna-Matthews-Olson light-harvesting antenna (FMO) protein has been a model system for understanding pigment-protein interactions in the energy transfer process in photosynthesis. All previous studies have utilized wild-type FMO proteins from several species. Here we report the purification and characterization
Chikara Ohto et al.
Applied microbiology and biotechnology, 87(4), 1327-1334 (2010-04-16)
An acyclic diterpene alcohol, (E,E,E)-geranylgeraniol (GGOH), is one of the important compounds used as perfume and pharmacological agents. A deficiency of squalene (SQ) synthase activity allows yeasts to accumulate an acyclic sesquiterpene alcohol, (E,E)-farnesol, in their cells. Since sterols are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service