Skip to Content
Merck
All Photos(1)

Key Documents

48809

Sigma-Aldrich

Geranyllinalool

technical, ≥90% (GC)

Synonym(s):

3,7,11,15-Tetramethyl-1,6,10,14-hexadecatetraen-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O
CAS Number:
Molecular Weight:
290.48
Beilstein:
6713421
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

refractive index

n20/D 1.490

density

0.885 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

C\C(C)=C/CC\C(C)=C\CC\C(C)=C\CCC(C)(O)C=C

InChI

1S/C20H34O/c1-7-20(6,21)16-10-15-19(5)14-9-13-18(4)12-8-11-17(2)3/h7,11,13,15,21H,1,8-10,12,14,16H2,2-6H3/b18-13+,19-15+

InChI key

IQDXAJNQKSIPGB-HQSZAHFGSA-N

General description

Geranyllinalool is a diterpene alcohol, commonly used as a fragrant. It imparts insecticidal property to xylophagous insects and pine wood.

Application

Geranyllinalool has been used in studying its influence on pyocyanin and Pseudomonas quinolone signal (PQS) production by Pseudomonas aeruginosa. It may be used as a starting material in the synthesis of teprenone by reacting with acetylated Meldrum′s acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diketene and Acetylated Meldrum's Acid: Agents for a Facile Teprenone Synthesis via Carroll Rearrangement.
De Castro KA, et al.
Bull. Korean Chem. Soc., 30(9), 2155-2157 (2009)
Jiao Xie et al.
Food chemistry, 243, 269-276 (2017-11-18)
Oleocellosis is a serious physiological disorder in citrus fruit that mainly results in appearance and quality deterioration. It has been well established that the occurrence of oleocellosis is highly correlated with the release of peel oil from citrus fruit, while
Sol A Green et al.
Journal of experimental botany, 63(5), 1951-1967 (2011-12-14)
Flowers of the kiwifruit species Actinidia chinensis produce a mixture of sesquiterpenes derived from farnesyl diphosphate (FDP) and monoterpenes derived from geranyl diphosphate (GDP). The tertiary sesquiterpene alcohol (E)-nerolidol was the major emitted volatile detected by headspace analysis. Contrastingly, in
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S176-S178 (2008-07-22)
A toxicologic and dermatologic review of geranyl linalool when used as a fragrance ingredient is presented.
Cinzia Formighieri et al.
Applied microbiology and biotechnology, 101(7), 2791-2800 (2017-01-08)
Cyanobacteria are industrially robust photosynthetic microorganisms that can be genetically programmed to synthesize commodity products for domestic and industrial consumption. In the present work, Synechocystis was endowed with the synthesis of the plant secondary metabolite geranyllinalool, a diterpene alcohol of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service