Skip to Content
Merck
All Photos(1)

Key Documents

277541

Sigma-Aldrich

trans,trans-Farnesol

96%

Synonym(s):

(E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol, trans,trans-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
222.37
Beilstein:
1723039
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.490 (lit.)

bp

137 °C/3 mmHg (lit.)

density

0.879 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CO

InChI

1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+

InChI key

CRDAMVZIKSXKFV-YFVJMOTDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans,trans-Farnesol is sesquiterpene alcohol, which is commonly found in many essential oils, citrus fruits, propolis, and plant-derived foods and beverages. It is an effective bioactive agent that can be used in pharmaceuticals and cosmetics.

Application

trans,trans-Farnesol has been used in the synthesis of:
  • Cecropia juvenile hormone
  • terpenes, such as squalenes, cembranoids and forskolin

Other Notes

A sesquiterpene alcohol found in many essential oils.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron Letters, 34, 1721-1721 (1993)
Assessment of the antioxidant and antiproliferative effects of sesquiterpenic compounds in in vitro Caco-2 cell models
Vinholes Juliana, et al.
Food Chemistry, 156, 204-211 (2014)
Influences of trans-trans farnesol, a membrane-targeting sesquiterpenoid, on Streptococcus mutans physiology and survival within mixed-species oral biofilms
Jeon J-G, et al.
International Journal of Oral Science, 3, 98-106 (2011)
Tetrahedron, 50, 139-139 (1994)
Tetrahedron, 49, 2457-2457 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service