Skip to Content
Merck
All Photos(4)

Key Documents

DUO92202

Sigma-Aldrich

Duolink® PLA Control Kit - PPI

Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA

Synonym(s):

in situ Proximity Ligation Assay Control Kit, Protein Protein Interaction Assay Control Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

product line

Duolink®

technique(s)

immunofluorescence: suitable
proximity ligation assay: suitable

suitability

suitable for fluorescence

shipped in

dry ice

storage temp.

−20°C

General description

Duolink® PLA Control Kits are intended for use with Duolink® PLA reagents to ensure proper function of reagents and Duolink® PLA staining technique.

Application

Application note

In the Duolink® PLA Control Kit, the cells and antibodies are provided, and the conditions have been optimized. When combined with the necessary Duolink® PLA reagents, you will have all you need to successfully perform the PLA experiment and obtain positive results.

The Control kit contains Duolink® PLA Control Slides (Cat. No. DUO82202) - two 8 well glass slides with fixed SK-OV3 Human ovarian cancer cells. The cells were treated with EGF, to induce EGFR and HER2 interaction. The Kit also contains one vial of Anti-HER2 antibody produced in rabbit (HPA001383), and one vial of Monoclonal Anti-EGFR antibody produced in mouse (E3138) – the two primary antibodies you need to detect the two interacting protein

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list
Based on proximity ligation assay (PLA), the Duolink® PLA Technology allows for endogenous detection of protein interactions, post-translational modifications, and protein expression levels at the single molecule level in fixed cells.

Duolink® PLA Control Kit - PPI is designed to allows users to gain confidence with the Duolink® PLA technology. This Kit contains glass slides with fixed, treated cells and a specific pair of primary antibodies that will detect interacting proteins and reliably generate PLA signals when used with the complementary Duolink® PLA probes and detection reagents. Analysis is carried out using standard immunofluorescence assay equipment.

Specificity
This kit will enable the detection of Protein-Protein interactions. The SK-OV-3 cells contain HER2 and EGFR, which show increased interaction upon treatment with EGF. When combined with appropriate Duolink® PLA Reagents, the Control Kit will enable the successful detection of increased HER2 and EGFR protein interactions in the EGF treated cells.

Features and Benefits

  • Reliable PLA signal detecting a protein interaction
  • Gain confidence with the Duolink® PLA Technology
  • Add additional controls to your PLA experiments
  • Increased sensitivity due to rolling circle amplification for low abundant targets
  • No overexpression or genetic manipulation required
  • Relative quantification possible
  • Works with any fluorescence microscope and appropriate filters

Components

This product is comprised of the following:
  • Duolink® PLA Control Slides (DUO82202) - two 8-well chamber slides with EGF treated, pre-fixed SK-OV3 cells.
  • One vial (15μL) of Rabbit anti-ErbB2/HER2 antibody (HPA001383)
  • One vial (25μL) of Mouse anti-EGFR antibody (E3138)

See datasheet for more information.

Other Notes

Other Duolink® reagents needed (30RXN size):
Mouse/Rabbit Duolink® PLA Starter Kit: Red Starter Kit (DUO92101) OR Orange Starter Kit (DUO92102)
OR
PLA Probe pairs (PLUS and MINUS): Anti-Mouse PLUS (DUO92001) and Anti-Rabbit MINUS (DUO92005) or Anti-Mouse MINUS (DUO92004) and Anti-Rabbit PLUS (DUO92002); AND Detection reagents of choice: Orange (DUO92007), Red (DUO92008), FarRed (DUO92013), or Green (DUO92014); AND Wash Buffers (DUO82049) AND Mounting Medium with DAPI (DUO82040).

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie Le Sage et al.
Virology, 502, 73-83 (2016-12-26)
Stress granules (SGs) are dynamic cytoplasmic aggregates of translationally silenced mRNAs that assemble in response to environmental stress. SGs appear to play an important role in antiviral innate immunity and many viruses have evolved to block or subvert SGs components
Franziska Wetzel et al.
Cellular and molecular life sciences : CMLS, 74(2), 373-392 (2016-09-09)
The zonula occludens (ZO)-2 protein links tight junctional transmembrane proteins to the actin cytoskeleton and associates with splicing and transcription factors in the nucleus. Multiple posttranslational modifications control the intracellular distribution of ZO-2. Here, we report that ZO-2 is a
Teng-Jian Zhou et al.
Theranostics, 7(5), 1389-1406 (2017-04-25)
Cancer stem cells (CSCs) are a small subset of malignant cells, possessing stemness, with strong tumorigenic capability, conferring resistance to therapy and leading to the relapse of nasopharyngeal carcinoma (NPC). Our previous study suggested that cyclooxygenase-2 (COX-2) would be a
Fredrick J Rosario et al.
The Journal of physiology, 595(13), 4189-4206 (2017-04-05)
Folate deficiency during pregnancy is associated with restricted fetal growth, although the underlying mechanisms are poorly understood. Here we show that mechanistic target of rapamycin (mTOR) functions as a folate sensor in primary human trophoblast (PHT) cells. Folate sensing by
Yujiao Lu et al.
Neurobiology of aging, 49, 165-182 (2016-11-07)
Beta amyloid (Aβ) is well accepted to play a central role in the pathogenesis of Alzheimer's disease (AD). The present work evaluated the therapeutic effects of low-level laser irradiation (LLI) on Aβ-induced neurotoxicity in rat hippocampus. Aβ 1-42 was injected

Articles

Learn how Proximity Ligation Assay technology works and how the protein-protein interaction control kit can confirm in situ detection of EGF-induced EGFR-HER2 dimerization.

Learn how Proximity Ligation Assay technology works and how the protein-protein interaction control kit can confirm in situ detection of EGF-induced EGFR-HER2 dimerization.

Protocols

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Related Content

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service