Skip to Content
Merck
All Photos(1)

Documents

D1529

Sigma-Aldrich

1-Fluoro-2,4-dinitrobenzene

≥99%

Synonym(s):

2,4-Dinitro-1-fluorobenzene, DNFB, DNPF, FDNB, Sanger reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3F
CAS Number:
Molecular Weight:
186.10
Beilstein:
398632
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥99%

form

solid

refractive index

n20/D 1.569 (lit.)

bp

178 °C/25 mmHg (lit.)

mp

25-27 °C (lit.)

density

1.482 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(F)c(c1)[N+]([O-])=O

InChI

1S/C6H3FN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H

InChI key

LOTKRQAVGJMPNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Fluoro-2,4-dinitrobenzene (DNFB) is a reagent that modifies the N-terminal amino acids of proteins and peptides. DNFB is also commonly known as Sanger′s Reagent, named after Frederick Sanger, who reported in a classic 1945 paper the use of DNFB for derivatization of proteins and peptides specifically at their N-termini.†

Application

1-Fluoro-2,4-dinitrobenzene has been used to induce severe contact hypersensitivity in mice and to monitor gene expression by in vivo imaging method, and to induce chronic itch as a model for analysing itch-scratching behavior.

Biochem/physiol Actions

1-Fluoro-2,4-dinitrobenzene is a contact-sensitizing hapten, commonly used in experimental studies on contact hypersensitivity, inducing itch model, and atopic dermatitis. Mast cell activation is a key step in the DNFB induced sensitivity. DNFB covalently binds to the N-terminal amino acid of the protein and aids in Sanger sequencing.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

327.2 °F

Flash Point(C)

164 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian C Palmer et al.
HSOA journal of toxicology : current research, 4(1) (2020-11-10)
Titanium dioxide (TiO2) nanoparticles are commonly found in consumer products, such as sunscreens, and human dermal exposures are relatively high. Research suggests potential differences in the toxicity of anatase and rutile crystalline forms of TiO2. Additionally, transition metal dopants are
Imaging protective mast cells in living mice during severe contact hypersensitivity
Reber LL, et al.
JCI insight, 2(9), e92900-e92900 (2017)
Bamboo salt suppresses skin inflammation in mice with 2, 4-dinitrofluorobenzene-induced atopic dermatitis
Yoou MS, et al.
Chinese Journal of Natural Medicines, 16(2), 97-104 (2018)
Beatrice Parma et al.
Journal of experimental & clinical cancer research : CR, 40(1), 248-248 (2021-08-09)
The identification of novel targets is of paramount importance to develop more effective drugs and improve the treatment of non-small cell lung cancer (NSCLC), the leading cause of cancer-related deaths worldwide. Since cells alter their metabolic rewiring during tumorigenesis and
Tac1-Expressing Neurons in the Periaqueductal Gray Facilitate the Itch-Scratching Cycle via Descending Regulation
Gao ZR, et al.
Neuron, 101(9), 45-59 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service