Skip to Content
Merck
All Photos(2)

Key Documents

C3029

Sigma-Aldrich

Potassium citrate tribasic monohydrate

suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5K3O7 · H2O
CAS Number:
Molecular Weight:
324.41
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (GC)

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

pH

8.0-9.5

solubility

H2O: 100 mg/mL, clear, colorless

SMILES string

O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3K.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

PJAHUDTUZRZBKM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clay Mechlin et al.
Journal of endourology, 25(9), 1541-1545 (2011-08-06)
Oral citrate supplements have been shown to decrease kidney stone recurrence rates in both laboratory and clinical studies. The taste of the citrate supplements, however, is poor, and long-term compliance is low. Our objective was to determine if Splenda(®) added
Chao-Yang Duan et al.
International journal of nanomedicine, 8, 909-918 (2013-03-08)
The property changes of urinary nanocrystallites in 13 patients with calcium oxalate (CaOx) stones were studied before and after ingestion of potassium citrate (K3cit), a therapeutic drug for stones. The analytical techniques included nanoparticle size analysis, transmission electron microscopy, X-ray
Hulya Nalcacioglu et al.
Journal of pediatric urology, 9(1), e58-e63 (2012-10-27)
Bilateral obstructive nephrolithiasis is a rare cause of acute kidney injury (AKI) in early childhood. As soon as the identification of AKI secondary to ureteral stone is made, it will necessitate an emergency treatment. We report three infants with AKI
Naim M Maalouf et al.
The Journal of clinical endocrinology and metabolism, 96(12), 3733-3740 (2011-10-07)
Dietary intake of animal proteins is associated with an increase in urinary calcium and nephrolithiasis risk. We tested the hypothesis that the acid load imposed by dietary proteins causes this hypercalciuria. In a short-term crossover metabolic study, an alkali salt
B Lojanapiwat et al.
International braz j urol : official journal of the Brazilian Society of Urology, 37(5), 611-616 (2011-11-22)
To evaluate the preventive effects of alkaline citrate on stone recurrence as well as stone growth post-ESWL or PCNL in patients with calcium-containing stones. A total of 76 patients with calcium calculi who were stone-free or had residual stones less

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service