Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium citrate tribasic monohydrate
Synonym(s):
Citric acid tripotassium salt, Tripotassium citrate
Linear Formula:
HOC(COOK)(CH
2
COOK)
2
· H
2
O
CAS Number:
6100-05-6
Molecular Weight:
324.41
EC Number:
231-905-0
Products
Product Category
buffers (3)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C8385
Potassium citrate tribasic monohydrate,
≥98% (GC/titration)
Expand
View Pricing
60153
Potassium citrate tribasic monohydrate,
purum p.a., ≥99.0% (NT)
Expand
View Pricing
C3029
Potassium citrate tribasic monohydrate,
suitable for cell culture
Expand
View Pricing
P1722
Potassium citrate tribasic monohydrate,
meets USP testing specifications
Expand
View Pricing
25107
Potassium citrate tribasic monohydrate,
meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Expand
View Pricing