Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
Porcine
Quality Segment
assay
≥99% (agarose gel electrophoresis)
form
lyophilized powder
technique(s)
cell culture | mammalian: suitable
UniProt accession no.
storage temp.
2-8°C
Gene Information
Porcine ... ALB(396960)
General description
Application
- as a non-hazardous protein to test its effect on T84 polarized intestinal monolayers and Caco2 polarized intestinal monolayers barrier integrity in trans-Epithelial Electrical Resistance (TEER) assay[2]
- to test its effect on the human intestinal epithelial cell lines[3]
- to test its interaction with the biodegradable ester-functionalized gemini surfactant 16-E2-16 using ultraviolet (UV)-visible and circular dichroism (CD) spectroscopy measurements
Biochem/physiol Actions
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| description lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis) | description lyophilized powder, 96-99% (agarose gel electrophoresis) | description lyophilized powder, ≥98% (agarose gel electrophoresis) | description lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis) |
| assay ≥99% (agarose gel electrophoresis) | assay 96-99% (agarose gel electrophoresis) | assay ≥98% (agarose gel electrophoresis) | assay ≥99% (agarose gel electrophoresis) |
| biological source Porcine | biological source Porcine | biological source Porcine | biological source rat |
| technique(s) cell culture | mammalian: suitable | technique(s) ELISA: suitable | technique(s) cell culture | mammalian: suitable | technique(s) ELISA: suitable, tissue culture: suitable, western blot: suitable |
| Quality Level 200 | Quality Level 200 | Quality Level 300 | Quality Level 200 |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| Gene Information Porcine ... ALB(396960) | Gene Information Porcine ... ALB(396960) | Gene Information Porcine ... ALB(396960) | Gene Information rat ... ALB(24186) |
Still not finding the right product?
Explore all of our products under Albumin from porcine serum
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



