Skip to Content
Merck

Skip To

A6414

Albumin from rat serum

lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)

Synonym(s):

RSA, Rat albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
rat
Assay:
≥99% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
ELISA: suitable, tissue culture: suitable, western blot: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

assay

≥99% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

rat ... ALB(24186)

General description

Albumin is the most abundant plasma protein in humans. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Albumin from rat serum has been used:
  • to study its sequence information from multiply charged ions formed by electrospray ionization
  • to cross-link with silk fibroin (SF) to synthesize a combined protein template
  • to infused into the brain to study its effects on nerve cell damage following brain injury

Preparation Note

Prepared from essentially globulin free albumin (A4538).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A4538A6272A3559
description

lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)

description

lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)

description

lyophilized powder, ≥96% (agarose gel electrophoresis)

description

lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

biological source

rat

biological source

rat

biological source

rat

biological source

mouse

technique(s)

ELISA: suitable, western blot: suitable, tissue culture: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

immunohistochemistry: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Albumin from rat serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I would like to know if the dried powder may contain traces of salts. More specifically, was it lyophilized from PBS, saline, salt solution, etc?

    1 answer
    1. Similar to product A3782, it was found that A6414 is dialyzed against water. However, there is no claim that ultrapure water was used. The available information simply states that the product was dialyzed against water. Additionally, there is no testing for salts based on the performed checks.

      Helpful?

Reviews

No rating value

Active Filters