Skip to Content

A1830

Albumin from porcine serum

lyophilized powder, ≥98% (agarose gel electrophoresis)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
Porcine
Assay:
≥98% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine

Quality Segment

assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

Porcine ... ALB(396960)

General description

Albumin from porcine serum (PSA) contains two tryptophan residues and amino Terminal Cu(II)- and Ni(II)-binding (ATCUN) motif with histidine-3 substituted by tyrosine-3 compared to human serum albumin.

Application

Albumin from porcine serum has been used in binding studies with:
  • Oxovanadium(IV)-O2+ complexes using circular dichroism (CD) and electron paramagnetic resonance studies
  • magnetic resonance imaging contrast agents, B229563 and MS-325 anion
  • ovine pregnancy-associated glycoprotein(PAG) radioimmunoassay (RIA) validation assay

Biochem/physiol Actions

Albumin from porcine serum (PSA) possesses foaming property and is an emulsifier. It is classified as an allergen and used in the preparation of molecularly imprinted nanogels for PSA detection on meat.


Still not finding the right product?

Explore all of our products under Albumin from porcine serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library