All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H24N6O9S3
Molecular Weight:
612.66
UNSPSC Code:
51282614
NACRES:
NA.85
Recommended Products
form
powder
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
Mode of action
cell wall synthesis | interferes
storage temp.
2-8°C
Related Categories
Biochem/physiol Actions
Cefpirome sulfate is a fourth generation cephalosporin antibiotic. It binds penicillin-binding proteins (PBPs) and inhibits bacterial cell wall synthesis.
Packaging
500mg
Other Notes
Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Hafeez et al.
JPMA. The Journal of the Pakistan Medical Association, 50(8), 250-252 (2000-09-19)
To study the in-vitro antimicrobial activity of Cefpirome: A new fourth generation Cephalosporin in comparison with other agents against clinically significant Gram-negative and Gram-positive bacteria. A multi-center in-vitro study was conducted in 13 centers. Bacterial isolates--A total of 1300 isolates
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service