Skip to Content
Merck
All Photos(1)

Key Documents

197491

Sigma-Aldrich

2,3-Cyclopentenopyridine

98%

Synonym(s):

6,7-Dihydro-5H-cyclopenta[b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N
CAS Number:
Molecular Weight:
119.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.544 (lit.)

bp

87-88 °C/11 mmHg (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

C1Cc2cccnc2C1

InChI

1S/C8H9N/c1-3-7-4-2-6-9-8(7)5-1/h2,4,6H,1,3,5H2

InChI key

KRNSYSYRLQDHDK-UHFFFAOYSA-N

General description

The structural properties of 2,3-cyclopentenopyridine using spectroscopic and computational techniques have been studied. 2,3-Cyclopentenopyridine is present in the side chain of cefpirome (HR 810), a new cephalosporin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdulaziz A Al-Saadi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 326-331 (2008-02-12)
The structural properties of 2,3-cyclopentenopyridine (pyrindan) have been investigated using several spectroscopic and computational techniques. The Raman and infrared spectra of the molecule have been recorded and a full vibrational assignment was proposed on the basis of experimental and theoretical
S Arai et al.
The Japanese journal of antibiotics, 43(1), 1-8 (1990-01-01)
Cefpirome (HR 810) is a new cephalosporin with a 2,3-cyclopentenopyridine group in the 3-position side chain. It was compared with other cephem antibiotics in protective and therapeutic effects on various experimental infections, systemic and local, in mice and rats. HR
Aldrichimica Acta, 11, 13-13 (1978)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service