Skip to Content
Merck

Skip To

77161

Pepsin from porcine gastric mucosa

tested according to Ph. Eur.

Synonym(s):

Pepsini pulvis, Pepsin A, Pepsin from hog stomach

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
42010127
EC Number:
232-629-3
NACRES:
NA.54
MDL number:
EC Number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

tested according to Ph. Eur.

form

powder

mol wt

35 kDa

UniProt accession no.

storage temp.

2-8°C

Gene Information

Application

Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.

Disclaimer

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7716077151P0609
description

tested according to Ph. Eur.

description

powder, slightly beige, ≥500 U/mg

description

powder, slightly beige, 1200-2400 U/mg

description

lyophilized powder, ≥30 units/mg dry solid

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

Gene Information

pig ... LOC396892(396892)

form

powder

form

powder

form

powder

form

lyophilized powder

UniProt accession no.

P00791

UniProt accession no.

P00791

UniProt accession no.

P00791

UniProt accession no.

P00791

mol wt

35 kDa

mol wt

35 kDa

mol wt

35 kDa

mol wt

35 kDa

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

agency

tested according to Ph. Eur.

agency

-

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Pepsin from porcine gastric mucosa


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hi, How many U/mL is in this product?

    1 answer
    1. This product is a powder with a minimum activity specification of 0.5 Ph. Eur. U/mg (calculated on dry substance). Kindly review the lot-specific certificate of analysis for the lot-specific result.

      Helpful?

Reviews

No rating value

Active Filters