Skip to Content
Merck
All Photos(1)

Documents

77151

Sigma-Aldrich

Pepsin from porcine gastric mucosa

powder, slightly beige, 1200-2400 U/mg

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010127
NACRES:
NA.54

form

powder

specific activity

1200-2400 U/mg

mol wt

35 kDa

color

slightly beige

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Unit Definition

1 U corresponds to the amount of enzyme which increases the absorbance at 280 nm by 0.001 per minute at pH 2.0 and 37°C (Hemoglobin, Cat.No. 51290, as substrate) 15′000 absorbance-U as described above are equivalent to ∼1 Bergmeyer-U. 1 Bergmeyer-U is the amount of enzyme which hydrolyzes 1 μmol Acetyl-L-phenyl-3,5-diiodo-L-tyrosine at pH 2.0 and 37°C

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.

Other Notes

Sales restrictions may apply

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhua Windt et al.
ACS omega, 7(44), 39924-39930 (2022-11-18)
Chicken feathers are major byproducts of the livestock processing industry with high potential in the feed sector. In this study, we present a new approach using Fourier transform infrared (FTIR) spectroscopy to detect the structural changes of feather keratin and
María Llarena-Reino et al.
Veterinary parasitology, 191(3-4), 276-283 (2012-10-10)
During the last 50 years human anisakiasis has been rising while parasites have increased their prevalence at determined fisheries becoming an emergent major public health problem. Although artificial enzymatic digestion procedure by CODEX (STAN 244-2004: standard for salted Atlantic herring
M N James et al.
Nature, 319(6048), 33-38 (1986-01-02)
The only well-understood mechanism of zymogen activation is that of the serine proteinases, in which proteolytic cleavage leads to conformational changes resulting in a functional active site. A different mechanism is now unveiled by the crystal structure of pepsinogen. Salt
Martial Rey et al.
Molecular & cellular proteomics : MCP, 12(2), 464-472 (2012-12-01)
Studies of protein dynamics, structure and interactions using hydrogen/deuterium exchange mass spectrometry (HDX-MS) have sharply increased over the past 5-10 years. The predominant technology requires fast digestion at pH 2-3 to retain deuterium label. Pepsin is used almost exclusively, but
Minghuo Wu et al.
Chemical communications (Cambridge, England), 49(14), 1407-1409 (2013-01-17)
A simple, novel "one-pot" approach was developed for the preparation of organic-silica hybrid monoliths. Two clickable monoliths functionalized with either azido or alkynyl moieties were prepared. The clickable monoliths were further used for the fabrication of enzyme reactors via "click

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service