Skip to Content
Merck
All Photos(1)

Documents

53134

Sigma-Aldrich

Streptavidin−Gold from Streptomyces avidinii

20 nm particle size

Synonym(s):

Streptavidin Gold conjugate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51102829
NACRES:
NA.25

form

liquid

Quality Level

particle size

20 nm

absorption

≤3.0 at 520 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For detection of biotinylated compounds.

Physical form

in 50 mM potassium phosphate, 75 mM NaCl buffer, pH 7.4 with 20% glycerol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service