85878
Streptavidin from Streptomyces avidinii
affinity purified, lyophilized from 10 mM potassium phosphate, ≥13 U/mg protein
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
powder
Quality Level
quality
affinity purified
lyophilized from 10 mM potassium phosphate
specific activity
≥13 U/mg protein
mol wt
Mr ~60000
solubility
H2O: 1 mg/mL, clear to hazy, colorless to faintly yellow
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Streptavidin is a crystalline protein synthesized by Streptomycetes.
Application
Streptavidin from Streptomyces avidinii has been used:
- in the pre-functionalization of self-propelled catalytic micromotors
- in the fabrication and modification of hydroxyapatite-chitosan (HA-CTS) nanofilm-coated substrates
- as an analyte to bind biomolecules to the gold-inverted polymer solar cells (Au-IPSC)
Biochem/physiol Actions
Streptavidin is a biotin binding protein and has the ability to bind four molecules of biotin. The biotin binding pocket of streptavidin helps to interact with biotin. It can be used to enhance protein binding and multimerization.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
The sales quantitites 1 mg and 5 mg
Unit Definition
1 U corresponds to the amount of protein which binds 1μg (+)-biotin at pH 7.5
Analysis Note
Binding capability: Streptavidin binds one molecule of biotin per subunit. Comparison of biotin binding and absorbance measurements indicates that ≤5% binding sites are occupied.
Other Notes
Review: Application of the avidin-biotin technique in microbiology; Protein isolated from the bacterium Streptomyces avidinii, having a high affinity for biotin; Avidin-Biotin technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nature communications, 10(1), 3907-3907 (2019-08-31)
Complementary assays are required to comprehensively map complex biological entities such as genomes, proteomes and interactome networks. However, how various assays can be optimally combined to approach completeness while maintaining high precision often remains unclear. Here, we propose a framework
Capture and identification of heterogeneous circulating tumor cells using transparent nanomaterials and quantum dots-based multiplexed imaging
Journal of Cancer, 7(1), 69-69 (2016)
Scientific and Clinical Applications of Magnetic Carriers (2013)
Evolved streptavidin mutants reveal key role of loop residue in high-affinity binding
Protein Science, 20(7), 1145-1154 (2011)
Biofunctionalized self-propelled micromotors as an alternative on-chip concentrating system
Lab on a chip, 14(16), 2914-2917 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service