Skip to Content

59758

Streptavidin particles, magnetic

≥2 U/mg

Synonym(s):

Magnetic streptavidin particles

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


specific activity

≥2 U/mg

concentration

5 mg/mL

storage temp.

2-8°C

Application

Streptavidin particles, magnetic may be used to bind biotin-conjugated molecules such as antibodies or DNA primers for use in gel agglutination assays.

Other Notes

1 U corresponds to the amount of particles which bind 1 ug (+)-biotin


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library