Skip to Content
Merck

Skip To

49101

β-Glucanase from Aspergillus niger

greener alternative

powder, dark brown, ~1 U/mg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-979-7
MDL number:
EC Number:
Specific activity:
~1 U/mg
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

specific activity

~1 U/mg

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

dark brown

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy eficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles.

Biochem/physiol Actions

β-glucanases degrade β-1,4-glucans of cellulose, xyloglucan and β-1,4-xylan.[1][2]

Other Notes

One unit corresponds to the amount of enzyme which will release 1 μmole of reducing sugar equivalents (expressed as glucose) per minute at pH 5.0 and 55 °C, using β-D-glucan (Cat. No. 49102) as substrate

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10113A7420A9857
specific activity

~1 U/mg

specific activity

~120 U/mg

specific activity

30-60 units/mg protein (biuret)

specific activity

≥150 units/mg protein (biuret)

form

powder

form

powder

form

lyophilized powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

color

dark brown

color

white

color

-

color

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

greener alternative category

Enabling,

greener alternative category

, Enabling

greener alternative category

, Enabling

greener alternative category

Enabling,


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I would like to know if this product digest beta 1-4 glucan or beta 1-3 glucan or both.

    1 answer
    1. As mentioned in the 'DESCRIPTION' section of the product page, under 'Biochem/physiol Actions', this product specifically degrades ?-1,4-glucans. Should a glucanase that degrades beta 1-3 glucan be required, please see product 67138 at the link below:
      https://www.sigmaaldrich.com/product/sigma/67138

      Helpful?

Reviews

No rating value

Active Filters