Skip to Content
Merck
All Photos(1)

Key Documents

380R-1

Sigma-Aldrich

Arginase-1 (SP156) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP156, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (380R-14)
vial of 0.5 mL concentrate (380R-15)
bottle of 1.0 mL predilute (380R-17)
vial of 1.0 mL concentrate (380R-16)
bottle of 7.0 mL predilute (380R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

hepatocellular carcinoma, normal liver

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, nuclear

Gene Information

human ... ARG1(383)

General description

Arginase is a key metalloenzyme of the urea cycle responsible for the hydrolysis of L-arginine to L-ornithine and urea. Two main isoforms exist, arginase-1 and arginase-2, encoded by different genes and with different tissue distributions. The arginase-1 isoform is a cytosolic protein that is produced by normal liver tissue and is typically expressed in hepatocellular carcinoma. Arginase-1 (SP156) is used as an immunohistochemical marker to aid in the identification of hepatocellular carcinoma.

Quality


IVD

IVD

IVD

RUO

Linkage

Arginase-1 Positive Control Slides, Product No. 380S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benign and malignant tumors of the liver
Linda DF
Surgical Pathology of the GI Tract, Liver, Biliary Tract, and Pancreas, 2nd ed., 1291-1325 (2009)
H B el-Serag
Clinics in liver disease, 5(1), 87-107 (2001-02-24)
The epidemiology of hepatocellular carcinoma (HCC) is characterized by marked differences between genders, ethnic groups, and geographic regions. These variations are explained by the nature, frequency, and time of acquisition of the major risk factors for cirrhosis--namely hepatitis B virus
Z W Zhu et al.
Gut, 48(4), 558-564 (2001-03-15)
Hepatocellular carcinoma (HCC) is a common malignant tumour worldwide, and its differential diagnosis from benign lesions of the liver is often difficult yet of great clinical importance. In the present study, we analysed whether glypican-3 is useful in differentiating between
Ahmedin Jemal et al.
CA: a cancer journal for clinicians, 61(2), 69-90 (2011-02-08)
The global burden of cancer continues to increase largely because of the aging and growth of the world population alongside an increasing adoption of cancer-causing behaviors, particularly smoking, in economically developing countries. Based on the GLOBOCAN 2008 estimates, about 12.7
H Multhaupt et al.
Histochemistry, 87(5), 465-470 (1987-01-01)
Monoclonal antibodies against human liver arginase were raised in order to determine the exact distribution of arginase in human liver using a modified indirect unlabelled immunoperoxidase method. In normal human liver specific immunohistochemical staining was found in the cytoplasm of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service