Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
15
biological source
chicken
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
guinea pig, human, mouse, rat
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgY
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ARG1(383)
mouse ... Arg1(11846)
rat ... Arg1(29221)
General description
Arginase-1, the protein encoded by the ARG1 gene, is the enzyme that catalyzes the breakdown of L-arginine into ornithine and urea. ARG1 is a key player in the Urea cycle. There are two distinct isoforms of Arginase, Arginase I and Arginase II. Arginase I (also called the liver type arginase or Type 1 arginase) is a highly expressed cytosolic enzyme in the liver and other tissues, including the brain. Arginase II or kidney-type arginase is also widely expressed at low levels in many tissues but localized to the mitochondria primarily. Interestingly, Arginase 1 has neuroprotective properties. Arginase I is essential for protecting motor neurons from nitric oxide dependent apoptosis that can be induced upon cellular stress or trophic factor removal. By suppressing nitric oxide production, Arginase prevents the formation of peroxynitrite and its subsequent cellular toxicity. Furthermore, Arginase 1 appears to play critical roles throughout the CNS for survival and regeneration of neurons and axons. In cancer, Arginase 1, already a useful marker for hepatocytes is also an excellent marker for hepatocellular cancers.
~ 38 kDa observed
Immunogen
Full-length recombinant protein corresponding to Rat Arginase-1.
Application
Immunohistochemistry Analysis: A representative lot detected Arginase-1 in bronchoalveolar lavage cells (Zimmermann, N., et al. (2003). J. Clin. Invest. 111:1863–1874 (2003)).
Western Blotting Analysis: A representative lot detected Arginase-1 in RAW cell lysate (Morris, S., et al. (1998). Am J Physiol Endocrinol Metab 275:E740-E747, 1998).
Western Blotting Analysis: A representative lot detected Arginase-1 in RAW cell lysate (Morris, S., et al. (1998). Am J Physiol Endocrinol Metab 275:E740-E747, 1998).
Research Category
Signaling
Signaling
This Anti-Arginase-1 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Arginase-1.
Physical form
Format: Purified
IgY purified from Yolk
Purified chicken Polyclonal other in buffer containing PBS with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in mouse LN tissue lysate.
Western Blotting Analysis: 2.0 µg/mL of this antibody detected Arginase-1 in 10 µg of mouse LN tissue lysate.
Western Blotting Analysis: 2.0 µg/mL of this antibody detected Arginase-1 in 10 µg of mouse LN tissue lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tim Tian Y Han et al.
Frontiers in bioengineering and biotechnology, 9, 642465-642465 (2021-04-06)
Cell-based therapies involving the delivery of adipose-derived stromal cells (ASCs) on decellularized adipose tissue (DAT) scaffolds are a promising approach for soft tissue augmentation and reconstruction. Our lab has recently shown that culturing human ASCs on DAT scaffolds within a
Daniella Bianchi-Frias et al.
Molecular cancer research : MCR, 17(1), 321-331 (2018-09-19)
The incidence of prostate cancer is directly linked to age, but age-associated changes that facilitate prostate cancer development and progression are poorly understood. This study investigated age-related changes in the prostate microenvironment for their influence on prostate cancer behavior. Prostate
Moaddey Alfarhan et al.
International journal of molecular sciences, 23(4) (2022-02-27)
Polyamine oxidation plays a major role in neurodegenerative diseases. Previous studies from our laboratory demonstrated that spermine oxidase (SMOX, a member of the polyamine oxidase family) inhibition using MDL 72527 reduced neurodegeneration in models of retinal excitotoxicity and diabetic retinopathy.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABS535 | 04053252988158 |