Skip to Content
Merck
All Photos(4)

Key Documents

C1016

Sigma-Aldrich

Calcium chloride

anhydrous, granular, ≤7.0 mm, ≥93.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
grade:
anhydrous
form:
granular

grade

anhydrous

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥93.0%

form

granular

particle size

≤7.0 mm

mp

772 °C (lit.)

storage temp.

room temp

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium chloride was used during the isolation of total β-glucan from hydrolyzed barley powders. It was also used as a cross-linker during the preparation of Aloe vera pectin gels.
It may be used in the following processes:
  • Modification of natural zeolite to increase its ability of fluoride removal.
  • Synthesis of calcium sulfate whiskers utilized in making polymer reinforcing composite materials.
  • Synthesis of calcium chloride-anodized aluminum composites, a solid sorbent.
  • As an extractant in the soil analysis process.
  • As a catalyst in the synthesis of aldols by reacting dimethylsilyl (DMS) enolates with aldehydes.

Caution

may contain powder and some dark grey granules

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Li et al.
Nano letters, 19(8), 5194-5204 (2019-07-02)
A globally imminent shortage of freshwater has been demanding viable strategies for improving desalination efficiencies with the adoption of cost- and energy-efficient membrane materials. The recently explored 2D transition metal dichalcogenides (2D TMDs) of near atomic thickness have been envisioned
Ángel Serrano-Aroca et al.
PloS one, 12(9), e0185235-e0185235 (2017-09-22)
Micrometer length tubes of graphene oxide (GO) with irregular form were synthesised following facile and green metal complexation reactions. These materials were obtained by crosslinking of GO with calcium, zinc or strontium chlorides at three different temperatures (24, 34 and
Sang Hoon Lee et al.
Preventive nutrition and food science, 20(2), 110-118 (2015-07-16)
This study was performed to investigate changes in the content and purity, as well as physical characteristics of β-glucan extracted from acid hydrolyzed whole grain barleys. Waxy and non-waxy barleys (Hordeum vulgare) were hydrolyzed with different concentrations of HCl (0.1~0.5
Pectins from Aloe Vera: Extraction and production of gels for regenerative medicine.
Gentilini R, et al.
Journal of Applied Polymer Science, 131(2) (2014)
Karine Glinel et al.
Biomacromolecules, 20(1), 102-108 (2018-07-07)
We demonstrate entrapment of the commensal skin bacteria Staphylococcus epidermidis in mats composed of soft nanotubes made by membrane-templated layer-by-layer (LbL) assembly. When cultured in broth, the resulting nanofibrillar patches efficiently delay the escape of bacteria and their planktonic growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service