Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium chloride
Synonym(s):
Calcium chloride
Linear Formula:
CaCl
2
CAS Number:
10043-52-4
Molecular Weight:
110.98
Products
Product Category
salts (13)
Brand
Sigma-Aldrich (11)
Supelco (5)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Anhydrous (12)
ACS reagent (2)
Purum (1)
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
C4901
Calcium chloride,
anhydrous, powder, ≥97%
Expand
View Pricing
C1016
Calcium chloride,
anhydrous, granular, ≤7.0 mm, ≥93.0%
Expand
View Pricing
C5670
Calcium chloride,
anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Expand
View Pricing
1.02378
Calcium chloride,
anhydrous granules Reag. Ph Eur
Expand
View Pricing
499609
Calcium chloride,
anhydrous, powder, 99.99% trace metals basis
Expand
View Pricing
383147
Calcium chloride,
desiccant, ACS reagent, ≥96.0%
Expand
View Pricing
746495
Calcium chloride,
anhydrous, free-flowing, Redi-Dri
™
, ≥97%
Expand
View Pricing
793639
Calcium chloride,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥96%
Expand
View Pricing
449709
Calcium chloride,
AnhydroBeads
™
, −10 mesh, ≥99.9% trace metals basis
Expand
View Pricing
21074
Calcium chloride,
purum, drying agent, dehydrated, granulated, ≥97.0% (KT)
Expand
View Pricing
1.02391
Calcium chloride,
anhydrous, granular ~ 2 - 6 mm
Expand
View Pricing
1.02379
Calcium chloride,
anhydrous, granular ~ 1-2 mm
Expand
View Pricing
1.02392
Calcium chloride,
anhydrous, granular ~ 6 - 14 mm
Expand
View Pricing
1.02083
Calcium chloride,
fused, granular about 0.5-2.0 mm for elementary analysis
Expand
View Pricing
429759
Calcium chloride,
AnhydroBeads
™
, −10 mesh, ≥99.99% trace metals basis
Expand
View Pricing
902179
Calcium chloride,
anhydrous, granular, ≤7.0 mm, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥93%
Expand
View Pricing