Skip to Content
Merck
All Photos(1)

Documents

21074

Sigma-Aldrich

Calcium chloride

purum, drying agent, dehydrated, granulated, ≥97.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

grade

drying agent
purum

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥97.0% (KT)

form

chunks or granules

quality

granulated

impurities

≤3% water

particle size

1-6 mm

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium chloride may be used for the preparation of self-supporting β-lactoglobulin gels. It was employed as an electrolyte to investigate the reduction of zirconium dioxide pellets by electro-deoxidation.

Caution

may contain powder and some dark grey granules

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical deoxidation of solid zirconium dioxide in molten calcium chloride.
Mohandas KS and Fray DJ.
Metall. Mater. Trans. B, 40(5), 685-699 (2009)
Gelation of β-Lactoglobulin: Effects of Sodium Chloride and Calcium Chloride on the Rheological and Structural Properties of Gels.Gelation of β-Lactoglobulin: Effects of Sodium Chloride and Calcium Chloride on the Rheological and Structural Properties of Gels.
Mulvihill DM and Kinsella JE.
Journal of Food Science, 53(1), 231-236 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service