Skip to Content
Merck
All Photos(3)

Key Documents

472476

Sigma-Aldrich

Chloroform

contains amylenes as stabilizer, ACS reagent, ≥99.8%

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.04
grade:
ACS reagent
Assay:
≥99.8%
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

contains

amylenes as stabilizer

impurities

Acetone and aldehyde, passes test
Acid and chloride, passes test
Free chlorine (Cl), passes test
≤0.03% (water)

evapn. residue

0.001%

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

cation traces

Pb: ≤0.05 ppm

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chloroform was used in the process for isolation of RNA from yeast.[1]
It may be used in the following processes:
  • Synthesis of colloidal hydrophobic CdSe core nanocrystals (quantum dots).[2]
  • Preparing phospholipid solutions.[3]
  • As a solvent in the photolysis reaction of ruthenium complexes.[4]
  • Solvent for recrystallization.[5]
  • Reference compound to detect the presence of chloroform in tobacco smokes.[6]
  • Extractant in solvent extraction process.[7]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple RNA Preparation Procedure from Yeast for Northern Blot Using Hot Phenol.
Wei Y.
Bio-protocol, 2(12) (2012)
Systematic studies on the breakdown of p,p'-DDT in tobacco smokes. Presence of methyl chloride, dichloromethane, and chloroform in tobacco smokes.
Chopra NM and Sherman LR.
Analytical Chemistry, 44(6), 1036-1038 (1972)
Engineering of ultra-small diagnostic nanoprobes through oriented conjugation of single-domain antibodies and quantum dots.
Sukhanova A, et al.
Protocol Exchange (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 216-216 (1994)
Kinetics and Mechanism of the Photooxidation of Bis (bipyridine) dichlororuthenium (II) and the Photoreduction of Bis (bipyridine) dichlororuthenium (III) in Chloroform.
Sathiyabalan S and Hoggard PE.
Inorganic Chemistry, 34(18), 4562-4571 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service