Skip to Content
Merck
All Photos(3)

Documents

178918

Sigma-Aldrich

Hexane, mixture of isomers

≥98.5%, ACS reagent

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

product name

Hexane, mixture of isomers, ACS reagent, ≥98.5%

grade

ACS reagent

Quality Level

vapor density

3 (vs air)

description

Mixture of hexane isomers plus methylcyclopentane

Assay

≥98.5%

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

impurities

thiophene, passes test
≤0.0003 meq/g water-soluble titr. acid
≤0.005% S compounds

evapn. residue

≤0.0010%

color

APHA: ≤10

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a mixture of hexane isomers and methylcyclopentane. Hexane (mixture of isomers) can be isolated from petroleum by distillation or by cracking. They form major constituents of petroleum ether. It is a non-renewable organic solvent mainly used for the dissolution and extraction of oils and fats. Separation of the isomeric forms of hexane on the basis of degree of branching using Fe2(BDP)3 (BDP2- = 1,4-benzenedipyrazolate) has been reported.

Application

Hexane (n-hexane) has been used in the preparation of precursor required for the synthesis of chiral stationary phase (IMPCSP). IMPCSP was employed for the enantioseparation of racemic drugs by high-performance liquid chromatography (HPLC) and supercritical fluid chromatography (SFC). It may be used as a carrier phase for microfluidic devices.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Copolymerization preparation of cationic cyclodextrin chiral stationary phases for drug enantioseparation in chromatography.
Wang RQ, et al.
Scientific Protocols null
Alternative bio-based solvents for extraction of fat and oils: solubility prediction, global yield, extraction kinetics, chemical composition and cost of manufacturing.
Sicaire A-G, et al.
International Journal of Molecular Sciences, 16(4), 8430-8453 (2015)
Solving the Clogging Problem: Precipitate-Forming Reactions in Flow.
Poe SL, et al.
Angewandte Chemie (International Edition in English), 45(10), 1544-1548 (2006)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Separation of hexane isomers in a metal-organic framework with triangular channels.
Herm ZR, et al.
Science, 340(6135), 960-964 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service