Skip to Content
Merck
All Photos(1)

Key Documents

650544

Sigma-Aldrich

Hexane, mixture of isomers

≥98.5%, HPLC Plus, for residue analysis, suitable for gas chromatography (GC) and HPLC

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.06
grade:
HPLC Plus
for residue analysis
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
bp:
68-70 °C (lit.)
Pricing and availability is not currently available.

Product Name

Hexane, mixture of isomers, HPLC Plus, for HPLC, GC, and residue analysis, ≥98.5%

grade

HPLC Plus
for residue analysis

vapor density

3 (vs air)

Assay

≥98.5%

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.01% water
≤1.0 ppb Fluorescence (quinine) at 365 nm

evapn. residue

≤0.0001%

halogenated residue

≤10 ng/L

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)

UV absorption

λ: 195 nm Amax: ≤1.00
λ: 225 nm Amax: ≤0.05
λ: 250 nm Amax: ≤0.01
λ: 275-300 nm Amax: ≤0.005

suitability

suitable for residue analysis

SMILES string

C(CCC)(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for HPLC, spectrophotometry, environmental testing

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service