Skip to Content
Merck
All Photos(2)

Key Documents

112615

Sigma-Aldrich

1-Octanol

ReagentPlus®, 99%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
196 °C (lit.)
vapor pressure:
0.14 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

4.5 (vs air)

Quality Level

vapor pressure

0.14 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

523 °F

expl. lim.

8 %

impurities

≤0.5% (water)

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

General description

1-Octanol (n-Octanol) is an ionic liquid[1] and a linear alcohol having an aromatic odor.It has a relatively high boiling point, low water solubility and moderate viscosity.[2] It can be synthesized from capric esters via catalytic hydrogenation.[3] Its oxidative decomposition in the presence of anchored photocatalysts (anchored titanium oxide catalysts and standard TiO2 powder) has been investigated.[4] Its liquid-phase oxidation to 1-octanal in the presence of Ti/Si binary oxide catalysts has been reported.[5] It has gained wide recognition as an extraction solvent in analytical chemistry.[1]

Application

1-Octanol may be used in the preparation of crystalline poly(vinylidene fluoride) (PVDF) membranes.[6]

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Application of ionic liquids in analytical chemistry"
Liu F-J, et al.
TrAC, Trends in Analytical Chemistry, 24(01), 20- 27 (2005)
"Single drop microextraction?development, applications and future trends"
Jeannot.AM, et al.
Journal of Chromatography A, 1217(16), 2326-2336 (2010)
Photocatalytic degradation of 1-octanol on anchored titanium oxide and on TiO2 powder catalysts.
Yamashita H, et al.
J. Catal., 158(1), 97-101 (1996)
Characterization of titanium-silicon binary oxide catalysts prepared by the sol-gel method and their photocatalytic reactivity for the liquid-phase oxidation of 1-octanol.
Yamashita H, et al.
The Journal of Physical Chemistry B, 102(30), 5870-5875 (1998)
Mechanisms of PVDF membrane formation by immersion-precipitation in soft (1-octanol) and harsh (water) nonsolvents.
Young TH, et al.
Polymer, 40(19), 5315-5323 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service