Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Octanol
Synonym(s):
Alcohol C
8
, Capryl alcohol, Octyl alcohol, 1-Octanol
Linear Formula:
CH
3
(CH
2
)
7
OH
CAS Number:
111-87-5
Molecular Weight:
130.23
EC Number:
203-917-6
Products
Product Category
solvents (4)
aroma chemicals (2)
Brand
Sigma-Aldrich (7)
Supelco (1)
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Kosher (2)
ACS reagent (1)
Anhydrous (1)
Food Grade (1)
Fragrance grade (1)
Halal (1)
Natural (1)
analytical standard (1)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
472328
1-Octanol,
ACS reagent, ≥99%
Expand
View Pricing
293245
1-Octanol,
suitable for HPLC, ≥99%
Expand
View Pricing
112615
1-Octanol,
ReagentPlus
®
, 99%
Expand
View Pricing
8.20931
1-Octanol,
for synthesis
Expand
View Pricing
W280003
1-Octanol,
≥98%, FCC, FG
Expand
View Pricing
297887
1-Octanol,
anhydrous, ≥99%
Expand
View Pricing
W280011
1-Octanol,
natural, ≥98%, FCC
Expand
View Pricing
95446
1-Octanol,
analytical standard
Expand
View Pricing