Skip to Content
Merck
All Photos(4)

Key Documents

03129

Sigma-Aldrich

Sodium iodide

puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.22

vapor density

>1 (vs air)

grade

puriss.

Assay

99-100.5% (calc. to the dried substance)

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

alkalic reac. Impurities, complies
barium and thiosulfate, complies
organic volatile impurities, complies (GC)
residual solvents, in accordance
≤0.0005% heavy metals (as Pb)
≤0.003% total nitrogen (N)
≤0.005% thiosulfate (S2O3)
≤0.02% free alkali (as NaOH)
≤2% water (Karl Fischer)

loss

≤2% loss on drying, 105 °C

pH

6-9 (20 °C, 50 g/L)

mp

661 °C (lit.)

anion traces

bromide, chloride (as Cl-): ≤300 mg/kg
iodate (IO3-): ≤2 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤5 mg/kg
K: ≤500 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].[I-]
[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium iodide is an inorganic salt. Mixture of chlorotrimethylsilane/sodium iodide in acetonitrile is a useful reagent for the cleavage of esters, lactones, carbamates and ethers.

Application

Sodium iodide may be used in the preparation of sodium hydroxide-sodium iodide reagent for the modified Winkler method for the determination of dissolved oxygen. It may be used for the synthesis of coppersensor-1 (CS1), a membrane-permeable fluorescent dye used for imaging labile copper pools in biological samples.

Caution

may contain black particle through the manufacture

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evan W Miller et al.
Nature protocols, 1(2), 824-827 (2007-04-05)
Coppersensor-1 (CS1) is a small-molecule, membrane-permeable fluorescent dye for imaging labile copper pools in biological samples, including live cells. This probe, comprising a boron dipyrromethene (BODIPY) chromophore coupled to a thioether-rich receptor, has a picomolar affinity for Cu+ with high
Synthetic methods and reactions. 62. Transformations with chlorotrimethylsilane/sodium iodide, a convenient in situ iodotrimethylsilane reagent.
Olah GA, et al.
The Journal of Organic Chemistry, 44(8), 1247-1251 (1979)
The Chesapeake Bay Institute technique for the Winkler dissolved oxygen method.
Carpenter JH.
Limnology and Oceanography, 10(1), 141-143 (1965)
Anke de Vries et al.
Investigative radiology, 50(4), 297-304 (2015-01-01)
The objective of this study was to investigate the feasibility and the accuracy of spectral computed tomography (spectral CT) to determine the tissue concentrations and localization of high-attenuation, iodine-based contrast agents in mice. Iodine tissue concentrations determined with spectral CT
Caroline Serrano-Nascimento et al.
Molecular and cellular endocrinology, 349(2), 154-161 (2011-10-18)
Iodide excess acutely downregulates NIS mRNA expression, as already demonstrated. PCCl3 cells treated or not with NaI, NaI+NaClO(4) or NaI+Methimazole, for 30 min to 24 h, were used to further explore how iodide reduces NIS gene expression. NIS mRNA expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service