Skip to Content
Merck
All Photos(1)

Key Documents

B95736

Sigma-Aldrich

Butyl isocyanate

98%

Synonym(s):

1-Isocyanatobutane, n-Butyl isocyanate, BIC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NCO
CAS Number:
Molecular Weight:
99.13
Beilstein:
773917
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3 (vs air)

Quality Level

vapor pressure

10.6 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

115 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCN=C=O

InChI

1S/C5H9NO/c1-2-3-4-6-5-7/h2-4H2,1H3

InChI key

HNHVTXYLRVGMHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Butyl isocyanate is a metabolite of benomyl, a broad spectrum fungicide.

Application

Butyl isocyanate can undergo anionic polymerization to form poly(butyl isocyanate) using lanthanum isopropoxide as the initiator.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Sik Yoon et al.
Environmental toxicology, 23(1), 131-144 (2008-01-25)
We investigated the toxic effects of carbendazim and n-butyl isocyanate (BIC), metabolites of the fungicide benomyl, on development in the African clawed frog, Xenopus laevis. To test the toxic effects, frog embryo teratogenesis assays using Xenopus were performed. Embryos were
J Pauluhn et al.
Archives of toxicology, 66(2), 118-125 (1992-01-01)
The objectives of this study were to use pulmonary function tests, blood gas measurements and bronchoalveolar lung lavage (BAL) to characterize lesions in the respiratory tract of young adult male Wistar rats as a result of a 5-day exposure (6
N, N′-dibutylurea from n-butyl isocyanate, a degradation product of benomyl. 1. Formation in Benlate formulations and on plants.
Moye H A, et al.
Journal of Agricultural and Food Chemistry, 42(5), 1204-1208 (1994)
Qinghai Huang et al.
Neurology India, 58(2), 259-263 (2010-05-29)
Distal anterior cerebral artery (DACA) aneurysms are less common, accounting for 3.1 to 9.2% of all intracranial aneurysms.The clinical characteristics and surgical techniques are quite different from those of other aneurysms of Willis circle. We aimed to investigate the clinical
S P Kucera et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 30(6), 779-799 (1995-11-01)
The effects of the fungicide benomyl and its breakdown products, carbendazim and butyl isocyanate, were examined on canine tracheal epithelial tissue in primary culture. Changes in ciliary frequencies were monitored with an optical spectrum analysis system. Serial dilutions of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service