1.06523
Sodium iodide
for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
Sodium iodide
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
vapor pressure
1 hPa ( 767 °C)
product line
EMSURE®
Assay
≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)
form
powder
potency
4340 mg/kg LD50, oral (Rat)
impurities
≤0.01% Insoluble matter
loss
≤ 0.5% loss on drying, 105 °C
pH
6-9 (20 °C, 50 g/L in H2O)
bp
1304 °C/1013 hPa
mp
659 °C
solubility
1793 g/L
density
3.67 g/cm3 at 20 °C
bulk density
1500‑2000 kg/m3
anion traces
chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤0.0003%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.002%
thiosulfate (S2O32-): passes test
cation traces
Ba: ≤0.002%
Ca: ≤0.001%
Cu: ≤0.0001%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.001%
N: ≤0.002%
Ni: ≤0.00001%
Pb: ≤0.0001%
heavy metals (as Pb): ≤0.0005%
storage temp.
2-30°C
SMILES string
[Na+].[I-]
InChI
1S/HI.Na/h1H;/q;+1/p-1
InChI key
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
- Sodium iodide symporter immunolabelling as a predictor of clinical iodide uptake in canine thyroid carcinoma: A preliminary study.: This research uses sodium iodide symporter (NIS) immunolabelling to predict iodide uptake in canine thyroid carcinoma, providing insights into the application of sodium iodide in cancer diagnostics and treatment (Dark et al., 2024).
- Photochemical Reactions in Dialdehyde Starch.: This article explores the photochemical reactions involving sodium iodide in dialdehyde starch, contributing to advancements in photochemistry and material science (Ziegler-Borowska et al., 2018).
Analysis Note
Assay (argentometric; calculated on dried substance): 99.5 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
Alkaline reacting substances: passes test
pH-value (5 %; water): 6.0 - 9.0
Chloride and Bromide (as Cl): ≤ 0.01 %
Iodate (IO₃): ≤ 0.0003 %
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.002 %
Thiosulfate (S₂O₃): passes test
Total nitrogen (N): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.001 %
Ni (Nickel): ≤ 0.00001 %
Pb (Lead): ≤ 0.0001 %
Loss on Drying (105 °C): ≤ 0.5 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral
Target Organs
Thyroid
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service