Skip to Content
Merck
All Photos(1)

Key Documents

Y0001525

Basic butylated methacrylate copolyme

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methyl-2-propenoic acid butyl ester polymer with 2-(dimethylamino)ethyl 2-methyl-2-propenoate and methyl 2-methyl-2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

copolymers, butylated methacrylate copolymer

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(CCOC(=O)C(=C)C)(C)C.O(CCCC)C(=O)C(=C)C.O(C)C(=O)C(=C)C

InChI

1S/C8H15NO2.C8H14O2.C5H8O2/c1-7(2)8(10)11-6-5-9(3)4;1-4-5-6-10-8(9)7(2)3;1-4(2)5(6)7-3/h1,5-6H2,2-4H3;2,4-6H2,1,3H3;1H2,2-3H3

InChI key

NEDGUIRITORSKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Basic butylated methacrylate copolyme EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Eisele et al.
Regulatory toxicology and pharmacology : RTP, 61(1), 32-43 (2011-06-28)
Basic Methacrylate Copolymer is a fully polymerised cationic copolymer with taste-masking and moisture protection properties. It is used as a pharmaceutical excipient and has potential use as a glazing/coating agent to solid food supplements. This article describes available information on
L S Ranzani et al.
Drug development and industrial pharmacy, 37(6), 694-701 (2011-01-14)
The aim of the present work was to investigate in vitro dissolution properties of three binary solid solutions prepared by a hot-melt extrusion (HME) process with vinyl pirrolidone--vinyl acetate copolymer (Kollidon VA 64), ethyl acrylate, methyl methacrylate polymer (Eudragit E)
Sheng Qi et al.
International journal of pharmaceutics, 354(1-2), 158-167 (2008-02-05)
Hot-melt extrusion has attracted considerable interest within the pharmaceutical industry. However, there remains some uncertainty as to how to characterise the physical structure of the extruded systems, particularly in terms of identifying the nature of the drug dispersion within the
Stefan Grube et al.
Biomacromolecules, 9(5), 1398-1405 (2008-04-15)
Membrane destabilizing properties and increased efflux of doxorubicin from liposomes caused by basic butylated methacrylate copolymer (BBMC), better known under its commercial trade name EUDRAGIT E, have been described in the scientific literature. Here, we investigated the effect of BBMC
A Sadato et al.
Academic radiology, 5(3), 198-206 (1998-04-02)
The authors evaluated the safety and efficacy of a nonadhesive liquid embolic material and the role of its components in histologic changes in embolized arteries. A Eudragit-E mixture (Rohm Chemische Fabrik, Darmstadt, Germany) was made of a cationic copolymer (Eudragit-E)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service