Skip to Content
Merck
All Photos(1)

Key Documents

Y0001487

Butyl methacrylate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COO(CH2)3CH3
CAS Number:
Molecular Weight:
142.20
Beilstein:
773960
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

4.91 (15 °C, vs air)

vapor pressure

2 mmHg ( 20 °C)

API family

butylated methacrylate copolymer

autoignition temp.

562 °F

expl. lim.

2-8 %

manufacturer/tradename

EDQM

refractive index

n20/D 1.423 (lit.)

bp

162-165 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCOC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Butyl methacrylate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Íris Duarte et al.
Pharmaceutical research, 32(1), 222-237 (2014-08-20)
To present a new screening methodology intended to be used in the early development of spray-dried amorphous solid dispersions. A model that combines thermodynamic, kinetic and manufacturing considerations was implemented to obtain estimates of the miscibility and phase behavior of
Makoto Ishida et al.
Drug development and industrial pharmacy, 40(8), 1054-1064 (2013-06-21)
Abstract The applicability of porous spherical silica (PSS) was evaluated as core particles for pharmaceutical products by comparing it with commercial core particles such as mannitol (NP-108), sucrose and microcrystalline cellulose spheres. We investigated the physical properties of core particles
Kateřina Punčochová et al.
International journal of pharmaceutics, 469(1), 159-167 (2014-04-23)
The objective of this study was to investigate the effect of different polymeric carriers in solid dispersions with an active pharmaceutical ingredient (API) on their water vapour sorption equilibria and the influence of the API-polymer interactions on the dissolution rate
Megan E Oest et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(3), 334-342 (2014-11-20)
Late-onset fragility fractures are a common complication following radiotherapy for metastatic disease and soft tissue sarcomas. Using a murine hindlimb focal irradiation model (RTx), we quantified time-dependent changes in osteoclasts and mineral apposition rate (MAR). Mice received either a single
Stanisław Popiel et al.
Analytica chimica acta, 837, 52-63 (2014-07-09)
Novel solid-phase microextraction (SPME) fibres containing methyl, ethyl, butyl acrylate and methacrylate were first prepared by a sol-gel technique and investigated for determination of selected organoarsenic compounds (lewisite, methyldichloroarsine, phenyldichloroarsine, diphenylchloroarsine and triphenylarsine) from water samples. The influence of sorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service