Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
leucovorin
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[Ca+2].[nH]1c2c([c](nc1N)=O)N(C(CN2)CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-])C=O
InChI
1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1
InChI key
KVUAALJSMIVURS-ZEDZUCNESA-L
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Folic Acid Tablets
- Leucovorin Calcium
- Leucovorin Calcium Compounded Oral Suspension
- Leucovorin Calcium for Injection
- Leucovorin Calcium Injection
- Leucovorin Calcium Tablets
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename EDQM | manufacturer/tradename EDQM |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format neat | format neat | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical |
| API family leucovorin | API family leucovorin | API family folic acid | API family - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
Still not finding the right product?
Explore all of our products under Leucovorin Calcium
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


