Skip to Content
Merck
All Photos(1)

Documents

PHR1420

Supelco

Doxylamine Succinate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Doxylamine succinate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O · C4H6O4
CAS Number:
Molecular Weight:
388.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001562
traceable to USP 1227006

API family

doxylamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)CCC(O)=O.CN(C)CCOC(C)(c1ccccc1)c2ccccn2

InChI

1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-9-5-4-6-10-15)16-11-7-8-12-18-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8)

InChI key

KBAUFVUYFNWQFM-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Doxylamine succinate is an antihistaminic drug generally employed for the prevention of morning sickness in pregnant women.

Application

Doxylamine Succinate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3445 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of a Rapid Chemometrics Assisted RP-HPLC with PDA Detection Method for the Simultaneous Estimation of Pyridoxine HCl and Doxylamine Succinate in Bulk and Pharmaceutical Dosage Form
Giriraj P and Sivakkumar T
Chromatography Research International, 2014(2), 91-97 (2014)
Liquid chromatographic determination of amines in complex cough-cold formulations
Fong GW and Mark EW
International Journal of Pharmaceutics, 53(2), 91-97 (1989)
Formulation and evaluation of orodispersible tablet of taste masked doxylamine succinate using ion exchange resin
Puttewar TY, et al.
Journal of King Saud University - Science, 22(4), 229-240 (2010)
Analysis of pharmaceutical preparations containing antihistamine drugs by micellar liquid chromatography
Martinez-Algaba C, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(2), 312-321 (2006)
Phillip Hasemann et al.
Electrophoresis, 28(11), 1779-1787 (2007-04-24)
An MEKC method was developed in order to separate a cold medicine formulation containing acetaminophen, ephedrine sulfate, doxylamine succinate, and dextromethorphan hydrobromide as active pharmaceutical ingredients. Because of their similar physical and chemical properties, it was a challenge to separate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service