
Doxylamine succinate

Synonym(s):
Doxylamine succinate salt
Empirical Formula (Hill Notation):
C17H22N2O · C4H6O4
CAS Number:
Molecular Weight:
388.46
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing