B14000
2-Benzoylpyridine
≥99%
Synonym(s):
Phenyl 2-pyridyl ketone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H9NO
CAS Number:
Molecular Weight:
183.21
Beilstein:
120283
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
form
crystals
bp
317 °C (lit.)
mp
41-43 °C (lit.)
SMILES string
O=C(c1ccccc1)c2ccccn2
InChI
1S/C12H9NO/c14-12(10-6-2-1-3-7-10)11-8-4-5-9-13-11/h1-9H
InChI key
GCSHUYKULREZSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yu-Lan Li et al.
Anti-cancer agents in medicinal chemistry, 20(4), 504-517 (2019-11-14)
To overcome the disadvantages of cisplatin, numerous platinum (Pt) complexes have been prepared. However, the anticancer activity and mechanism of Pt(II) complexed with 2-benzoylpyridine [Pt(II)- Bpy]: [PtCl2(DMSO)L] (DMSO = dimethyl sulfoxide, L = 2-benzoylpyridine) in cancer cells remain unknown. Pt(II)-Bpy
Ahmed S M Al-Janabi et al.
Journal of molecular structure, 1228, 129454-129454 (2020-10-27)
New Schiff bases {N'-(phenyl(pyridin-2-yl)methylene) isonicotinohydrazide (L1H), N1 -(naphthalen-1-yl)-N2 -(phenyl(pyridin-2-yl) methylidene) ethane-1,2-diamine (L2H), N-(6-chlorobenzo[d]thiazol-2-yl)-1-phenyl-1-(pyridin-2-yl) methanimine (L3H)}were synthesized by reaction of 2-benzoylpyridine with different amines (2-amino-6-chlorobenzothiazole, isonicotinohydrazide and N 1-(naphthalen-1-yl)ethane-1,2-diamine) and characterized by 1H-NMR, 13C-NMR, IR mass spectroscopy and elemental analysis. The
Sandeep Kumar Dash et al.
Journal of applied toxicology : JAT, 34(11), 1130-1144 (2014-01-31)
The aim of the present study was to develop zinc sulfide nanoparticles (ZnS NPs) and to study their cytotoxicity against the KG-1A (human acute myeloid leukemia) cell line. ZnS NPs were synthesized using the pyrolytic method and characterized by X-ray
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service