Skip to Content
Merck
All Photos(2)

Key Documents

41868

Supelco

Ginsenoside Rb2

analytical standard

Synonym(s):

(3β,12β)-20-[(6-O-α-L-Arabinopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, NSC 308878

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C/CC[C@](C)(O[C@@H]1O[C@H](CO[C@@H]2OC[C@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)[C@H]3CC[C@]4(C)[C@@H]3[C@H](O)C[C@@H]5[C@@]6(C)CC[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)C(C)(C)[C@@H]6CC[C@@]45C

InChI

1S/C53H90O22/c1-23(2)10-9-14-53(8,75-47-43(67)39(63)37(61)29(72-47)22-69-45-41(65)34(58)26(57)21-68-45)24-11-16-52(7)33(24)25(56)18-31-50(5)15-13-32(49(3,4)30(50)12-17-51(31,52)6)73-48-44(40(64)36(60)28(20-55)71-48)74-46-42(66)38(62)35(59)27(19-54)70-46/h10,24-48,54-67H,9,11-22H2,1-8H3/t24-,25+,26-,27+,28+,29+,30-,31+,32-,33-,34-,35+,36+,37+,38-,39-,40-,41+,42+,43+,44+,45-,46-,47-,48-,50-,51+,52+,53-/m0/s1

InChI key

NODILNFGTFIURN-GZPRDHCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Hu Quan et al.
Journal of industrial microbiology & biotechnology, 39(10), 1557-1562 (2012-06-22)
The ginsenoside-hydrolyzing β-glycosidase (Bgp3) derived from Microbacterium esteraromaticum transformed the major ginsenoside Rb2 to more pharmacologically active minor ginsenosides including compounds Y and K. The bgp3 gene consists of 2,271 bp encoding 756 amino acids which have homology to the
Fang Liu et al.
Chemical & pharmaceutical bulletin, 56(3), 389-393 (2008-03-04)
In this paper, a new method for liquid chromatographic fingerprint of saponins in Gynostemma pentaphyllum (THUNB.) MAKINO was developed. The G. pentaphyllum powder was defatted by Soxhlet extraction with petroleum ether and then gypenosides were extracted from the residue with
Huarong Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(2), 312-316 (2007-02-28)
The noncovalent complexes of cytochrome c and ginsenoside were studied by electrospray ionization mass spectrometry (ESI-MS). Ginsenoside Rb2 and Re were bound to cytochrome c to form several complexes with different stoichiometric relation. The 1:1 and 1:2 complexes of cytochrome
E A Bae et al.
Biological & pharmaceutical bulletin, 23(12), 1481-1485 (2001-01-06)
When ginsenoside Rb1 and Rb2 were anaerobically incubated with human intestinal microflora, these ginsenosides were metabolized to 20-O-beta-D-glucopyranosyl-20(S)-protopanaxadiol (compound K) and 20(S)-protopanaxadiol. Several kinds of intestinal bacteria hydrolyzed these ginsenosides. Eubacterium sp., Streptococcus sp. and Bifidobacterium sp., which more potently
Kyoung-Tae Lee et al.
Archives of pharmacal research, 34(7), 1201-1208 (2011-08-04)
Ginsenosides, which are active compounds found in ginseng (Panax ginseng), are used as antidiabetic treatments. The aim of this study was to determine whether Rb2, a type of ginsenoside, regulates hepatic gluconeogenesis through AMP-activated protein kinase (AMPK) and the orphan

Articles

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service