Skip to Content
Merck
All Photos(1)

Key Documents

33434

Supelco

Ciprofloxacin

VETRANAL®, analytical standard

Synonym(s):

1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, Ciprobay

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O3
CAS Number:
Molecular Weight:
331.34
Beilstein:
3568352
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

InChI

1S/C17H18FN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)

InChI key

MYSWGUAQZAJSOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: fluoroquinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orjan Samuelsen et al.
Antimicrobial agents and chemotherapy, 54(1), 346-352 (2009-11-04)
Scandinavia is considered a region with a low prevalence of antimicrobial resistance. However, the number of multidrug-resistant (MDR) Gram-negative bacteria is increasing, including metallo-beta-lactamase (MBL)-producing Pseudomonas aeruginosa. In this study MBL-producing P. aeruginosa isolates identified in Norway (n = 4)
Elena B M Breidenstein et al.
Antimicrobial agents and chemotherapy, 52(12), 4486-4491 (2008-10-01)
Pseudomonas aeruginosa offers substantial therapeutic challenges due to its high intrinsic resistance to many antibiotics and its propensity to develop mutational and/or adaptive resistance. The PA14 comprehensive mutant library was screened for mutants exhibiting either two- to eightfold increased susceptibilities
Hong Bin Kim et al.
Antimicrobial agents and chemotherapy, 53(8), 3582-3584 (2009-06-17)
The genes for multidrug efflux pump OqxAB, which is active on fluoroquinolones, were found in human clinical isolates on a plasmid in Escherichia coli and on the chromosome of Klebsiella pneumoniae. IS26-like sequences flanked the plasmid-mediated oqxAB genes, suggesting that
Motohide Sato et al.
Journal of medicinal chemistry, 49(5), 1506-1508 (2006-03-03)
The viral enzyme integrase is essential for the replication of human immunodeficiency virus type 1 (HIV-1) and represents a remaining target for antiretroviral drugs. Here, we describe the modification of a quinolone antibiotic to produce the novel integrase inhibitor JTK-303
Li-Yang Hsu et al.
Antimicrobial agents and chemotherapy, 54(3), 1173-1178 (2010-01-13)
A surveillance study was performed in four Singapore public hospitals from 2006 to 2008 to determine the correlation between antibiotic prescription and Gram-negative bacterial antimicrobial resistance. Targeted organisms included ceftriaxone- and ciprofloxacin-resistant Escherichia coli and Klebsiella pneumoniae, as well as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service