Skip to Content
Merck
All Photos(1)

Documents

Y0000198

Ciprofloxacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, Ciprobay

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O3
CAS Number:
Molecular Weight:
331.34
Beilstein:
3568352
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciprofloxacin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

InChI

1S/C17H18FN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)

InChI key

MYSWGUAQZAJSOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciprofloxacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Rodríguez-Pardo et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 20(11), O911-O919 (2014-04-29)
We aim to evaluate the epidemiology and outcome of gram-negative prosthetic joint infection (GN-PJI) treated with debridement, antibiotics and implant retention (DAIR), identify factors predictive of failure, and determine the impact of ciprofloxacin use on prognosis. We performed a retrospective
Florentia Kaguelidou et al.
The Pediatric infectious disease journal, 30(2), e29-e37 (2010-11-05)
ciprofloxacin has no marketing authorization for use in neonates worldwide but it is prescribed for the treatment of neonatal life-threatening infections, mainly in developing countries and in Europe. Given the concerns about its toxicity in this population and the necessity
Mark D P Willcox
Clinical & experimental optometry, 94(2), 161-168 (2010-11-19)
Microbial keratitis is a rare disease but most commonly caused by bacterial infection. Two of the most common bacteria to cause microbial keratitis are Pseudomonas aeruginosa and Staphylococcus aureus. Antibiotic therapy to treat keratitis caused by these bacteria is either
Elisabet Marti et al.
Water research, 61, 67-76 (2014-06-02)
In this study we characterised the ciprofloxacin-resistant strains isolated in biofilm and sediments from a wastewater treatment plant (WWTP) discharge point and its receiving river. We also examined the prevalence of qnrA, qnrB, qnrS and aac(6')-Ib-cr genes in these isolates
David J Serisier et al.
Thorax, 68(9), 812-817 (2013-05-18)
The delivery of antipseudomonal antibiotics by inhalation to Pseudomonas aeruginosa-infected subjects with non-cystic fibrosis (CF) bronchiectasis is a logical extension of treatment strategies successfully developed in CF bronchiectasis. Dual release ciprofloxacin for inhalation (DRCFI) contains liposomal ciprofloxacin, formulated to optimise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service