Skip to Content
Merck
All Photos(1)

Key Documents

34031

Supelco

Aflatoxin M1 solution

0.5 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O7
CAS Number:
Molecular Weight:
328.27
Beilstein:
1630643
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

COc1cc2OC3OC=C[C@@]3(O)c2c4OC(=O)C5=C(CCC5=O)c14

InChI

1S/C17H12O7/c1-21-9-6-10-13(17(20)4-5-22-16(17)23-10)14-12(9)7-2-3-8(18)11(7)15(19)24-14/h4-6,16,20H,2-3H2,1H3/t16-,17-/m1/s1

InChI key

MJBWDEQAUQTVKK-IAGOWNOFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial
Fungal genera like Aspergillus, Fusarium, Penicillium, etc., produce mycotoxins. These toxic secondary metabolites are harmful to animal and human health if they enter the food chain via contaminated crops and fruits or through processed food and feed products.
Aflatoxin M1 is a single-component mycotoxin standard solution developed for precise quality control of food and feed.
Isoproturon-d6 has been used as an internal standard in the analysis of pesticides in fine airborne particulate matter, using liquid extraction under pressure and liquid chromatographic–tandem mass spectrometric method (HPLC-MS-MS).

Application

Aflatoxin M1 may be used as an analytical reference standard for the quantification of the analyte in milk samples and milk products using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography/tandem mass spectrometric confirmatory method for determining aflatoxin M1 in cow milk: comparison between electrospray and atmospheric pressure photoionization sources.
Cavaliere C, et al.
Journal of Chromatography A, 1101(1-2), 69-78 (2006)
"Analysis of currently used pesticides in fine airborne particulate matter (PM 2.5) by pressurized liquid extraction and liquid chromatography?tandem mass spectrometry"
Coscolla C, et al.
Journal of Chromatography A, 1200(02), 100-107 (2008)
Determination of Aflatoxin M1 in Milk and Milk Powder Using High-Flow Solid-Phase Extraction and Liquid Chromatography- Tandem Mass Spectrometry
Chen Y-C, et al.
Journal of Agricultural and Food Chemistry, 53(22), 8474-8480 (2005)
High-performance liquid chromatography with post-column derivatisation and fluorescence detection for sensitive determination of aflatoxin M1 in milk and cheese.
Manetta AC, et al.
Journal of Chromatography A, 1083(1-2), 219-222 (2005)
Sabran Mohd Redzwan et al.
Bulletin of environmental contamination and toxicology, 89(6), 1115-1119 (2012-10-12)
This study aimed to find the association between urinary aflatoxin M(1) level and milk and dairy products consumption. Of 160 morning urine samples collected, aflatoxin M(1) was detected in 61.3 % samples (n = 98) [mean ± SD = 0.0234

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service