Skip to Content
Merck
All Photos(1)

Key Documents

30494

Supelco

Silydianin

analytical standard

Synonym(s):

Silidianin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% residual solvents (GC-HS)
≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(ccc1O)[C@@H]2[C@H]3CO[C@]4(O)[C@H]3C(=C[C@H]2C4=O)[C@H]5Oc6cc(O)cc(O)c6C(=O)[C@@H]5O

InChI

1S/C25H22O10/c1-33-16-4-9(2-3-14(16)27)18-11-7-12(20-13(18)8-34-25(20,32)24(11)31)23-22(30)21(29)19-15(28)5-10(26)6-17(19)35-23/h2-7,11,13,18,20,22-23,26-28,30,32H,8H2,1H3/t11-,13-,18+,20+,22+,23-,25-/m1/s1

InChI key

CYGIJEJDYJOUAN-JSGXPVSSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Małgorzata Zielińska-Przyjemska et al.
Phytotherapy research : PTR, 20(2), 115-119 (2006-01-31)
The inhibitory effect of silydianin, an active constituent of Silybium marianum, on the in vitro production and release of oxidative products has been examined. Polymorphonuclear neutrophils (PMNs) play a primary role in the initiation and propagation of inflammatory responses. Their
János Fehér et al.
Orvosi hetilap, 149(51), 2413-2418 (2008-12-17)
In the treatment of chronic liver diseases adequate therapy can be chosen only in the knowledge of pathogenetic processes. In the liver diseases caused by oxidative stress (alcoholic and non-alcoholic fatty liver and steatohepatitis, drug and compound induced liver toxicity)
Cheng Lu et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 66(2), 210-219 (2007-01-02)
Development of sustained delivery systems for herbal medicines was very difficult because of their complexity in composition. The concept of synchronized release from sustained release systems, which is characterized by release of multiple components in their original ratio that defines
[Bioavailability of silymarin, I: Volumes of distribution of silybin, silydianin, and silychristin from in-vitro data (author's transl)].
H P Koch et al.
Archiv der Pharmazie, 314(6), 515-517 (1981-06-01)
Roman Zuber et al.
Phytotherapy research : PTR, 16(7), 632-638 (2002-11-01)
Silybin and related flavonolignans form a major part of the Silybum marianum extract, silymarin, which has been used to treat liver diseases for hundreds of years. Although regarded as safe, many of the extract constituents remain thus far untested for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service