
Silydianin

Synonym(s):
Silidianin, Silydianin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
Products
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3