Skip to Content
Merck
All Photos(4)

Key Documents

296082

Sigma-Aldrich

Diethyl ether

≥99.7%, anhydrous, contains 1 ppm BHT as inhibitor

Synonym(s):

DEE, Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99.7%
bp:
34.6 °C (lit.)
34.6 °C
vapor pressure:
28.66 psi ( 55 °C)
8.56 psi ( 20 °C)
Pricing and availability is not currently available.

Product Name

Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%

grade

anhydrous

Quality Level

vapor density

2.6 (vs air)

vapor pressure

28.66 psi ( 55 °C)
8.56 psi ( 20 °C)

Assay

≥99.7%

form

liquid

autoignition temp.

320 °F

contains

1 ppm BHT as inhibitor

expl. lim.

36.5 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

color

colorless

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)
34.6 °C

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl ether is a highly volatile and flammable organic solvent.[1] On storing for a long time, DEE can form organic peroxides, which can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT scavenges the free radicals required for the peroxide formation.

Application

Diethyl ether has been used as a solvent for the preparation of 3-formyl-2,2,5,5-tetramethyl-1-oxy-pyrroline from 3-(N-methoxy-N-methylcarbamoyl)-2,2,5,5-tetramethyl-1-oxy-pyrroline using DIBAL.[2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Merck Index: An Encyclopedia of Chemicals, Drugs,
Royal Society of Chemistry null
Site-directed spin-labeling of nucleotides and the use of in-cell EPR to determine long-range distances in a biologically relevant environment
Azarkh M, et al.
Nature Protocols, 8(1), 131-131 (2013)
Diethyl ether (DEE) as a renewable diesel fuel.
No. 972978. SAE technical paper (1997)
Marta Guembe-García et al.
Polymers, 12(6) (2020-06-04)
We anchored a colourimetric probe, comprising a complex containing copper (Cu(II)) and a dye, to a polymer matrix obtaining film-shaped chemosensors with induced selectivity toward glycine. This sensory material is exploited in the selectivity detection of glycine in complex mixtures
Haijun Tong et al.
International journal of nanomedicine, 8, 1935-1946 (2013-06-06)
Genetically modified mesenchymal stem cells (MSCs) have great potential in the application of regenerative medicine and molecular therapy. In the present manuscript, we introduce a nanopolymer, polyethylenimine600-β-cyclodextrin (PEI600-β-CyD), as an efficient polyplex-forming plasmid delivery agent with low toxicity and ideal

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Could you please advise of the shelf life of 296082

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service