Skip to Content
Merck
All Photos(1)

Key Documents

Z677736

Vitoject fluorocarbon rubber gloves

size XL (11)

Synonym(s):

chemically resistant gloves, hand protection against aggressive chemicals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.

material

black (fluorocarbon rubber)
black gloves

size

XL (11)

sterility

non-sterile

feature

CE compliant
latex free
powder-free: no
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)

packaging

pack of 1 pair

manufacturer/tradename

KCL 890

L × thickness

350 mm × 0.7 mm

Looking for similar products? Visit Product Comparison Guide

General description

Vitoject is made of 100% high chemical resistant Viton®, absolutely indispensable when working with aromatic hydrocarbons (Benzene, Toluene, ...), chlorinated hydrocarbons (Tri, Per, ...), fuels, aggressive acids and many other chemicals.

Manufactured with the ecologically beneficial and modern Vulcoject-process technology (injection molding).

  • Very good protection against extremely aggressive chemicals
  • 100% gastight
  • Excellent wearing comfort
  • Solvent free by eco-friendly injection molding process
  • CE compliant

Legal Information

Vitoject is a trademark of KCL GmbH
Viton is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service