Skip to Content
Merck
All Photos(1)

Key Documents

17777

Millipore

Potassium chlorate Supplement

suitable for microbiology

Synonym(s):

Potassium chlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Agency

according to ISO 10273:2017

Quality Level

sterility

sterile

form

liquid

shelf life

limited shelf life, expiry date on the label

mp

356 °C (lit.)

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Yersinia spp.

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A liqiud Potassium chlorate supplement for the selective enrichment of Yersinia enterococlitica.

Components

(per flask, sufficient for 1000 ml medium)
Potassium chlorate 1 g
distilled water 30 ml

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Bosnjak et al.
American journal of physiology. Heart and circulatory physiology, 284(4), H1080-H1086 (2003-02-22)
Thrombin (Thromb), activated as part of the clotting cascade, dilates conduit arteries through an endothelial pertussis toxin (PTX)-sensitive G-protein receptor and releases nitric oxide (NO). Thromb also acts on downstream microvessels. Therefore, we examined whether Thromb dilates human coronary arterioles
Paola González Audino et al.
Pest management science, 58(2), 183-189 (2002-02-21)
The stability to heart of cis-permethrin and beta-cypermethrin in the solid phase was studied and the decomposition products identified. Samples heated at 210 degrees C in an oven in the dark showed that, in the absence of potassium chlorate (the
Mónica Millán et al.
International journal of pharmaceutics, 310(1-2), 168-174 (2006-01-25)
The main objective of this work is to study the influence of the drug particle size on the pharmaceutical availability of ultrasound compacted tablets. Inert matrix systems containing different drug particle sizes were prepared using both, an ultrasound-assisted press and
Kazutomo Ishi et al.
Bioscience, biotechnology, and biochemistry, 69(12), 2463-2465 (2005-12-27)
The nitrate reductase gene (niaD) is the most frequently utilized as a selectable marker for homologous integration at the niaD locus of Aspergillus oryzae. In this study we developed a method for curing of the niaD-based plasmid integrated on the
Sang Jin Lee et al.
Journal of biomaterials science. Polymer edition, 13(2), 197-212 (2002-05-23)
It has been recognized that adhesion and proliferation of cells on biodegradable polymers such as poly(lactic acid) (PLA), poly(glycolic acid) (PGA), and poly(lactide-co-glycolide) (PLGA) depend on the surface properties. The chloric acid (CA) treatment of these films was developed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service