Skip to Content
Merck
All Photos(2)

Key Documents

1.05043

Supelco

Potassium iodide

for analysis EMSURE® ISO,Reag. Ph Eur

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-659-4
NACRES:
NA.21
form:
solid
solubility:
1430 g/L
Pricing and availability is not currently available.

Agency

reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 745 °C)

product line

EMSURE®

Assay

≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)

form

solid

potency

2779 mg/kg LD50, oral (Rat)

impurities

≤0.001% Total nitrogen (N)

loss

≤0.1% loss on drying, 105 °C

pH

6.9 (20 °C, 50 g/L in H2O)

bp

1325 °C/1013 hPa

mp

681 °C (lit.)

solubility

1430 g/L

bulk density

1500 kg/m3

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤0.0003%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.001%
thiosulfate (S2O32-): passes test

cation traces

As: ≤0.00001%
Ba: ≤0.002%
Ca: ≤0.001%
Cu: ≤0.0002%
Fe: ≤0.0002%
Mg: ≤0.001%
N: ≤0.001%
Na: ≤0.03%
Pb: ≤0.0002%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Influence of silver fluoride plus potassium iodine on adhesive properties of primary carious dentin-resin interface: An in vitro study.: This study examines how potassium iodide, in combination with silver fluoride, impacts the bonding properties of dental resin to carious dentin, highlighting its potential in dental treatments (da Luz et al., 2024).
  • Comparisons of ammonia- and water-based silver-containing solutions on dentin bonding and enzymatic activity: 1-yr evaluation.: The research investigates the effects of potassium iodide in silver-containing solutions on dentin bonding and enzyme activity, providing insights into dental material performance over time (D′ Alessandro et al., 2024).
  • Multi-spectroscopic and in silico investigation of gambogic acid-calf thymus DNA interactions.: This study utilizes potassium iodide in spectroscopic methods to elucidate the interactions between gambogic acid and DNA, contributing to the understanding of its potential therapeutic effects (Bhardwaj et al., 2024).
  • Surface Modification of Lignite with Alkyl and Mixed Alkyl-Aryl Films Generated from an Aryl Diazonium Salt and Alkyl Halides: Experimental Results and Theoretical Analyses.: Research details the use of potassium iodide in the surface modification of lignite, presenting a blend of experimental and theoretical insights to improve material properties (Hasani et al., 2024).

Analysis Note

Assay (argentometric): ≥ 99.5 %
Assay (argentometric; calculated on dried substance): 99.5 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Alkaline impurities: passes test
pH-value (5 %; water): 6 - 8
Chloride and Bromide (as Cl): ≤ 0.01 %
Iodate (IO₃): ≤ 0.0003 %
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.001 %
Thiosulfate (S₂O₃): passes test
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.00001 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0002 %
Mg (Magnesium): ≤ 0.001 %
Na (Sodium): ≤ 0.03 %
Pb (Lead): ≤ 0.0002 %
Reducing substances: passes test
Loss on Drying (105 °C): ≤ 0.1 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

See All

Protocols

Preparation of a standard solution for Chlorine dioxide acc. to DIN EN ISO 7393

Silica gel G 254 plates are suitable for analysis of Dextromethorphan following the European pharmacopeia monograph.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service