Skip to Content
Merck
All Photos(1)

Key Documents

17192

Millipore

Peptone Sorbitol Bile Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74
Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bile salt mixture, 1.5 g/L
disodium phosphate, 8.23 g/L
monosodium phosphate, 1.2 g/L
peptic digest of animal tissue, 5 g/L
sodium chloride, 5 g/L
sorbitol, 10 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.6±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

selective for Yersinia spp.

Application

For identification of Yersinia enterocolitica from dairy products.

Preparation Note

Suspend 30.93 g in 1 litre distilled water. Heat to boiling to dissolve the medium completely. Dispense 100 ml into Wheaton bottles. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M F Peruzy et al.
International journal of food microbiology, 301, 51-60 (2019-05-18)
Wild boars (Sus scrofa) are the most widely distributed large mammals and recent increase in consumption of wild boar meat urges the need of microbiological quality criteria. The aim of the study was to characterize the initial bacterial contamination on
Maria Francesca Peruzy et al.
International journal of food microbiology, 336, 108902-108902 (2020-10-23)
Routine evaluation of the slaughter process is performed by the enumeration of the aerobic colony count, Enterobacteriaceae and Salmonella spp. on the carcass through destructive or non-destructive methods. With non-destructive methods, bacteria are counted from a minimum area of 100 cm2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service