Skip to Content
Merck
All Photos(1)

Documents

15209

Supelco

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane

for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)

Synonym(s):

BSTFA + TMCS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Beilstein:
2050024
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥97.5% (excluding TMCS, GC)
98% (excluding TMCS)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

impurities

9.0-11.0% TMCS

refractive index

n20/D 1.384 (lit.)

bp

45-50 °C/14 mmHg (lit.)

density

0.97 g/mL (lit.)

SMILES string

C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+

InChI key

XCOBLONWWXQEBS-KPKJPENVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane has found application to be used in gas chromatography (GC) and mass-spectrometry (MS). It can also be used in HPLC- electrochemical detection assay of 8-oxo-deoxyguanosine and?8-oxo-guanine.

Application


  • Determination of Δ(9)-tetrahydrocannabinol, 11-nor-carboxy-Δ(9)-tetrahydrocannabinol and cannabidiol in human plasma and urine after a commercial cannabidiol oil product intake.: This study uses N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for derivatization to enhance the sensitivity and accuracy of cannabinoid detection in biological samples. This method is vital for forensic toxicology and clinical research, providing reliable data for cannabinoid analysis (Papoutsis et al., 2024).

  • The impact of lipid degradation on fingerprint quality on fired firearm cartridges.: This research employs N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane to study lipid degradation and its effect on fingerprint quality on firearm cartridges. The findings are crucial for forensic science, particularly in improving methods for preserving and analyzing fingerprint evidence (Goulart et al., 2023).

  • Contaminants of emerging concern: Silylation procedures, evaluation of the stability of silyl derivatives and associated measurement uncertainty.: This paper evaluates the use of N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane in silylation procedures for environmental analysis. The study assesses the stability and measurement uncertainty of silyl derivatives, which is crucial for accurate monitoring of environmental contaminants (Ljoncheva et al., 2023).

  • Screening of Organic Acidurias by Gas Chromatography-Mass Spectrometry (GC-MS).: This research utilizes N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane in the derivatization process for screening organic acidurias. This method is significant for clinical diagnostics, providing a reliable approach for detecting metabolic disorders (Scott et al., 2022).

  • A sensitive, robust method for determining natural and synthetic hormones in surface and wastewaters by continuous solid-phase extraction-gas chromatography-mass spectrometry.: The study presents a method using N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for the detection of hormones in environmental samples. This robust method is essential for monitoring water quality and detecting endocrine-disrupting compounds (Chafi and Ballesteros, 2022).


Other Notes

The position of the silyl groups has not been unequivocally established. It has been referred to as both N,N and N,O.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Metabolism of arachidonate through NADPH-dependent oxygenase of renal cortex.
Proceedings of the National Academy of Sciences of the USA, 78 (12), 7375-7378 (1981)
DNA oxidation matters: the HPLC-electrochemical detection assay of 8-oxo-deoxyguanosine and 8-oxo-guanine.
Helbock H
Proceedings of the National Academy of Sciences of the USA, 95 (1), 288-293 (1998)
Hidenori Matsukami et al.
The Science of the total environment, 493, 672-681 (2014-07-06)
Chlorinated and non-halogenated organophosphorus flame retardants (OPFRs) including tris(2-chloroisopropyl) phosphate (TCIPP), diethylene glycol bis(di(2-chloroisopropyl) phosphate) (DEG-BDCIPP), triphenyl phosphate (TPHP), and bisphenol A bis(diphenyl phosphate) (BPA-BDPP) have been used increasingly as alternatives to polybrominated diphenyl ethers and other brominated flame retardants.
Minghua Nie et al.
The Science of the total environment, 487, 785-791 (2013-12-25)
The occurrence and multi-phase distribution of six environmental estrogen compounds were investigated in a drinking water reservoir area by analyzing estrogens in suspended particulate matter (SPM), filtrate (conventional dissolved phase, <1 μm), permeate (truly soluble phase, <1 kDa) and retentate
Laura Degani et al.
Analytical and bioanalytical chemistry, 407(6), 1695-1704 (2015-01-19)
The possibility of successfully applying a common GC-MS procedure for identification in one step of all types of dyes from plants of unknown origin and from historical objects is particularly attractive due to the high separation efficiency of the capillary

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service