Merck
  • Environmental estrogens in a drinking water reservoir area in Shanghai: occurrence, colloidal contribution and risk assessment.
Environmental estrogens in a drinking water reservoir area in Shanghai: occurrence, colloidal contribution and risk assessment.
The Science of the total environment (2013-12-25)
Minghua Nie, Yi Yang, Min Liu, Caixia Yan, Hao Shi, Wenbo Dong, John L Zhou
ABSTRACT
The occurrence and multi-phase distribution of six environmental estrogen compounds were investigated in a drinking water reservoir area by analyzing estrogens in suspended particulate matter (SPM), filtrate (conventional dissolved phase, <1 μm), permeate (truly soluble phase, <1 kDa) and retentate (colloidal phase, 1 kDa to 1 μm). The estrogen concentrations at different sites occurred in the following order: animal feed operation (AFO) wastewater-affected streams>tributaries>main stream channel. Correlation analysis showed that organic carbon (OC) contents had significantly positive correlations with environmental estrogens in filtrate, SPM and colloidal phases, respectively, indicating the important role played by OC. Aquatic colloids, often neglected, showed a much higher sorption capability of environmental estrogens compared to SPM. Similar Kcoc values in three types of sampling sites showed that colloids could be transported from AFO wastewater to tributaries and further into the main river channel. Mass balance calculations showed that 14.5-68.4% of OP, 4.5-32.1% of BPA, 2.0-58.4% of E1, 8.36-72.0% of E2, 0-20.6% of EE2, 3.4-62.7% of E3 and 8.3-36.1% of total estrogens were associated with colloidal fractions, suggesting that the colloids could act as a significant sink for environmental estrogens. Risk assessment demonstrated that the occurrence of environmental estrogens might pose a risk to aquatic organisms in the study area.
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, LiChropur
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
Chlorotrimethylsilane, LiChropur, ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
Metolachlor OA, PESTANAL®, analytical standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Trifluoroacetamide, 97%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, LiChropur, ≥99.0%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%